missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Vivaspin™ 6 Centrifugal Concentrators

Achieve fast sample concentration and high recovery. Sartorius™ Vivaspin™ 6 Centrifugal Concentrators are the only device of its size that can process 6mL samples. They feature twin vertical membranes and built-in dead stop pockets. Available with PES membranes in a wide range of molecular weight cut-offs.

Brand:  Sartorius™ VS06S1

Code : 53

Additional Details : Weight : 2.14100kg

 View more versions of this product

Product Code. 10738421

  • £161.00 / Pack of 25


Estimated Shipment
23-03-2018
Add to basket

Description

Description

Developed to offer increased volume flexibility and performance
  • Compact tubes can process up to an impressive 6mL initial volume
  • Optimally suited for the concentration of antibody, protein, nanoparticle and virus samples
  • Universal rotor compatibility:
    • Use in either swing bucket or fixed angle rotors accepting standard 15mL conical bottom test tubes
Fast, safe and easy concentration
  • Features premium, high-throughput PES membranes
  • Twin vertical membrane design lead to unparalleled filtration speed and over 100-fold concentration
  • The dead stop pocket prevents concentration to dryness for safe sample concentration
  • Nearly complete concentrate recovery of over 90%
  • Remaining volume is easy to see with the printed graduations and clear housing
  • Modified dead stop pocket further simplifies direct pipette recovery of the final concentrate
Specifications

Specifications

Polyethersulfone
122mm
25/Pk.
6mL
Polycarbonate
Starter Pack: 5 of each 5K, 10K, 30K, 50K, 100K
2.58cm2
Videos
Documentation