missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Vivapure™ miniprep A / G spin columns

Centrifugal units with Protein A or Protein G resin for fast affinity purification of antibodies from small samples up to 0.65 ml.

Brand:  Sartorius™ VS-ARSTPKA2

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11770845

  • £125.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Vivapure miniprep and spin columns are pre-packed centrifugal devices for purification of monoclonal and polyclonal antibodies from serum, ascites and cell culture supernatants. The rapid 1-2-3 bind-wash-elute protocol especially lends itself to screening applications, where many samples are processed in parallel. Vivapure miniprep & spin columns can conveniently be used in a centrifuge with swing-out rotor for standard 2.2 ml tubes. Binding capacity is about 1 mg IgG per unit. The low hold-up volume ensures high solute recovery with minimal loss by non-specific adsorption. Vivapure miniprep and spin columns can be re-used three times and are available as complete kits including buffers or as bulk packs.They are an attractive alternative to lengthy chromatographic methods such as FPLC. For larger volumes, the Vivapure Maxiprep kit is also availablity.

  • Spin column based kit
  • Devices are ready to use
  • Rapid purification with high yields
  • Parallel processing
  • Cost effective through re-use option
  • Starter kits available

Specifications

Specifications

Purification of Monoclonal and Polyclonal Antibodies from Serum, ascites and cell culture supernatants
MiniprepA starter pack
3 Users
UF-concentrato rs and Buffers
2 Pack
Videos