missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Vivacon™ 2 DNA Concentrator

Centrifugal units for fast DNA concentration and buffer exchange of 2 ml samples

Brand:  Sartorius™ VN02H33

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11770846

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

45 min.
4.92 x 0.62 in. (12.5 x 1.6cm)
11.5cm
Polycarbonate Body, Polypropylene filtrate vessel, Polypropylene back spin vial, Polypropylene concentrator cap, Hydrosart™ membrane, Silicone O-ring
50000 MWCO
DNA, peptide
1.6cm
Vivacon™ 2 DNA Concentrator
2mL
50ng/mL
0.95cm2
12.5cm
2500xG
300 bp
10μL
DNA concentration, desalting, buffer exchange, fractionation
500 Pack