missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Vivacon™ 2 DNA Concentrator

Centrifugal units for fast DNA concentration and buffer exchange of 2 ml samples

Brand:  Sartorius™ VN02H93

UNSPSC : 41120000

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11790846

  • £1839.31 / Pack of 500


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

120 min.
4.92 x 0.62 in. (12.5 x 1.6cm)
11.5cm
Polycarbonate Body, Polypropylene filtrate vessel, Polypropylene back spin vial, Polypropylene concentrator cap, Hydrosart™ membrane, Silicone O-ring
2000 MWCO
DNA, peptide
1.6cm
Vivacon™ 2 DNA Concentrator
2mL
50ng/mL
0.95cm2
12.5cm
7500xG
10 bp
10μL
DNA concentration, desalting, buffer exchange, fractionation
500 Pack