Learn More
Sartorius™ Stedim Biotech Ink Jet Printer
Used for arium™ models.
Brand: Sartorius™ 611APR1
Code : NEW
Additional Details : Weight : 10.00000kg
Description
- Pre-programmed and supplied with data and power cables
- Can be connected to the arium in a matter of minutes
- Date, time, resistance or conductivity will be automatically sent to the printer at time intervals as selected
- No further manipulation of data is required
Specifications
Ink Jet Printer | |
480 mm | |
100/240 V |
430 mm | |
334 mm | |
50/60 Hz |