Learn More
Sartorius Sartolab™ RF/BT Vacuum Filtration Units
Filter aqueous solutions quickly and easily. Sartorius Sartolab™ RF/BT Vacuum Filtration Units are disposable and easy to use filter units. They contain a high quality PES membrane and combine the highest flow-rates and throughput with extremely low protein binding and extractables. Available in a range of volumes.
Brand: Sartorius 180C3----------E
Code : 53
Additional Details : Weight : 3.91500kg
Description
Quality, reliable sterile filtration units- Receiver Flask (RF) and Bottle Top (BT) units contain a high quality Sartorius PES membrane
- Combine the highest flow-rates and throughput with extremely low protein binding and extractables
- Ideal for filtration of tissue culture media, buffers, additives and other aqueous biologicals
- RF and BT vacuum units are available as 150mL, 250mL, 500mL and 1000mL units
- 500mL and 1000mL units come standard with 90mm membranes
- Improved filtration speed and throughput for cost effective filtration
Specifications
USP Class VI, ISO 10993 | |
Single-use | |
Polystyrene | |
0.22 μm | |
62 cm2 | |
12 Case | |
1000 mL |
Plug-on hose nipple | |
1.77 in. bottle neck | |
PES | |
Sterile | |
Filtration of tissue culture media, buffers, additives and other aqueous biologicals | |
Sartolab RF Filter |