missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Sartolab™ RF/BT Vacuum Filtration Units

Filter aqueous solutions quickly and easily. Sartorius™ Sartolab™ RF/BT Vacuum Filtration Units are disposable and easy to use filter units. They contain a high quality PES membrane and combine the highest flow-rates and throughput with extremely low protein binding and extractables. Available in a range of volumes.

Brand:  Sartorius™ 180C2----------E

Code : 53

Additional Details : Weight : 6.35940kg

 View more versions of this product

Product Code. 11781005

  • £207.00 / Pack of 12


In Stock 4
Add to basket

Description

Description

Quality, reliable sterile filtration units
  • Receiver Flask (RF) and Bottle Top (BT) units contain a high quality Sartorius PES membrane
  • Combine the highest flow-rates and throughput with extremely low protein binding and extractables
  • Ideal for filtration of tissue culture media, buffers, additives and other aqueous biologicals
  • RF and BT vacuum units are available as 150mL, 250mL, 500mL and 1000mL units
    • 500mL and 1000mL units come standard with 90mm membranes
  • Improved filtration speed and throughput for cost effective filtration
Specifications

Specifications

USP Class VI, ISO 10993
Yes
39cm2
1.77 in. bottle neck
Sartolab RF Filter
0.22μm
Yes
Plug-on hose nipple
Polyethersulfone
Filtration of tissue culture media, buffers, additives and other aqueous biologicals
Polystyrene
500mL
12 Case
Videos
Documentation