missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Quantitative Grade 392 Filter Papers
Brand: Sartorius™ FT-3-105-125
Code : NEW
Additional Details : Weight : 0.10000kg
Description
- Ash-free (ash content ≤ 0.01 % according to DIN 540370)
- Wet-strengthened
- Color-coded box for easy selection
Specifications
0.17mm | |
392 | |
125mm | |
Red |
100 per pack | |
Disc | |
5 to 8μmm |