missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Quantitative Grade 392 Filter Papers

Brand:  Sartorius™ FT-3-105-185

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10429322

  • £50.20 / Pack of 100


Estimated Shipment
23-02-2018
Add to basket

Description

Description

  • Ash-free (ash content ≤ 0.01 % according to DIN 540370)
  • Wet-strengthened
  • Color-coded box for easy selection
Specifications

Specifications

0.17mm
392
185mm
Red
100 per pack
Disc
5 to 8μmm
Videos