missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Quantitative Grade 392 Filter Papers

Brand:  Sartorius FT-3-105-110

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10130661

Quantity Price
1 £24.26 / Pack of 100
10 £22.65 / Pack of 100
Estimated Shipment: 23-11-2021

Add to basket


Description

Description

  • Ash-free (ash content ≤ 0.01 % according to DIN 540370)
  • Wet-strengthened
  • Color-coded box for easy selection
Specifications

Specifications

Red
Disc
5 to 8μm
0.17 mm
110 mm
392
100/Pk.
Videos
SDS
Documents
Certificates