missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Quantitative Grade 390 Filter Papers

Brand:  Sartorius™ FT-4-103-125

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11701385

  • £34.80 / Pack of 100


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Ash-free (ash content ≤ 0.01 % according to DIN 540370)
  • Wet-strengthened
  • Color-coded box for easy selection
Specifications

Specifications

100 per pack
Folded
1 to 2μmm
0.18mm
390
320mm
Green
Videos