missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Quantitative Grade 292 Filter Papers

Brand:  Sartorius™ FT-3-205-125

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10307261

  • £11.60 / Pack of 100


Estimated Shipment
15-02-2018
Add to basket

Description

Description

  • Pure cotton linters or cotton linters with refined pulp
  • No additives, such as wet-strengthening agents
Specifications

Specifications

0.18mm
292
125mm
100 per pack
Disc
5 to 8μmm
Videos