missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Quantitative Grade 292 Filter Papers
These filter papers are especially suitable for qualitative analyses that require a low ash content.
£8.10 - £212.00
Specifications
Format | Disc |
---|---|
Particle Retention | 5 to 8 μm |
Filtration Speed | 60 sec. |
Grade | 292 |
Quantity | 100/Pk. |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10215801
|
Sartorius
FT-3-205-185 |
185 mm |
£36.30
Pack of 100 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
15952890
|
Sartorius
FT-3-205-175 |
175 mm |
£33.80
Pack of 100 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
10377351
|
Sartorius
FT-3-205-320 |
320 mm |
£62.40
Pack of 100 |
In Stock
Log in to see stock. |
|||||
15962890
|
Sartorius
FT-3-205-270 |
270 mm |
£59.00
Pack of 100 |
In Stock
Log in to see stock. |
|||||
15972890
|
Sartorius
FT-3-205-500 |
500 mm |
£212.00
Pack of 100 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
10194271
|
Sartorius
FT-3-205-240 |
240 mm |
£59.00
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10565321
|
Sartorius
FT-3-205-150 |
150 mm |
£24.20
Pack of 100 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
10307261
|
Sartorius
FT-3-205-125 |
125 mm |
£24.10
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10632392
|
Sartorius
FT-3-205-055 |
55 mm |
£10.20
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10524571
|
Sartorius
FT-3-205-070 |
70 mm |
£13.05
Pack of 100 |
In Stock
Log in to see stock. |
|||||
10594751
|
Sartorius
FT-3-205-090 |
90 mm |
£11.10
Pack of 100 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
10114181
|
Sartorius
FT-3-205-110 |
110 mm |
£14.50
Pack of 100 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
10721563
|
Sartorius
FT-3-205-030 |
30 mm |
£8.10
Pack of 100 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
10086971
|
Sartorius
FT-3-205-042 |
42 mm |
£8.10
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
Description
- Pure cotton linters or cotton linters with refined pulp
- No additives, such as wet-strengthening agents
Specifications
Disc | |
60 sec. | |
100/Pk. |
5 to 8 μm | |
292 | |
0.18 mm |