Sartorius™ Minisart™ HY Syringe Filters

Designed for fast and reliable sterile venting and gas filtration. Sartorius™ Minisart™ HY Syringe Filters are single-use, complete filtration units with a naturally hydrophobic PTFE membrane and medical acrylic housing. Choose from 0.2 and 1.0µm pore sizes. Sterile option available.

Overview
Brand: Sartorius™

Manufacturer Part Number: 1659B--------HYQ

UNSPSC: 41120000

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11700816

Quantity Price
1 £ 718.0 / Pack of 500
Estimated Shipment
11-04-2017
Add to basket


Description and Specification

Specification

Inlet Style Female Luer Lok
Filtration Area (Metric) 5.16cm2
Housing Material Medical acrylic (MBS)
Color Clear
Filter Type Hydrophobic
Hold up Volume (After Air Purge) 0.25mL
Disposable Yes
Pressure Rating (Metric) 4.5bar
Outlet Style Male Luer Lok
Material Polytetrafluoroethylene (PTFE)
Diameter (Metric) 26mm
Type Syringe Filter
Sterile No
Quantity 500/pk.
Tip Material Cryolite
Pore Size 1μm

Fast and reliable sterile venting and filtration
  • Clean, safe filters that are almost free of leachables and extractables
  • Reliably removes particles and microorganisms with no leakage
  • Has minimal effect on the ingredients of the filtered solution
  • Includes a naturally hydrophobic, 26mm PTFE membrane with medical acrylic (MBS) housing
  • Wide variety of lab uses:
    • Reliable filtration of particulates and cells from gasses, additives, buffers and reagents
    • Sterile venting of small fermenters or bioreactors and of containers for culture media
    • Also used for aqueous and oily ear- or eye-solutions and other drugs
Convenient and economical
  • Complete units are ready to use
  • Single use/disposable filter units
  • Female luer lock inlet connectors with male leur lock outlet connector
  • Proven quality and 100% tested during manufacture
  • Sterile (via ethylene oxide) and non-sterile units available