Learn More
Sartorius Microsart™ e.motion Membrane Filters
For colony counting, particle testing and microscopy. Specifically designed for use in the Microsart™ e.motion dispenser, the filters are individually sterile-sealed in a format packaged on a pleated band, without protective paper. Can also be used with Sartorius nutrient pad sets.
Brand: Sartorius 114H6Z-47----SCM
Additional Details : Weight : 2.00000kg
Description
For colony counting, particle testing and microscopy. Specifically designed for use in the Microsart™ e.motion dispenser, the filters are individually sterile-sealed in a format packaged on a pleated band, without protective paper. Can also be used with Sartorius nutrient pad sets.
Microsart™ e.motion Membrane Filters, 0.45μm HIGH FLOW, white/black, 47 mm. The Microsart™ e.motion membrane filters are suitable for the use with the Microsart™ e.motion dispenser. The filters are individually sterile-sealed on a band without any protective paper. The special pleating as well as the shape of the sealing of the band of membrane filters ensures a perfectly flat and uniform dispensing of the individual membrane filters. The Microsart™ e.motion membrane filters are also ideal for all users of the Sartorius Nutrient Pad Sets: most of the NPS types are available with Microsart™ e.motion membrane filters. Outstanding recovery rates for microorganisms 0.45 μm are acc. to ISO 7704. Multi-fit: Fits into other dispensers. Protective paper-free. Packaged on a special pleated band. Product data is printed on individual envelopes. High flow membranes. Gamma irradiated, 25 kGray.Specifications
White | |
Cellulose Nitrate | |
0.45 μm | |
For colony count; sterility testing; particle count; microscopy | |
300/Pk. | |
47 mm |
Membrane Filter | |
Cellulose Nitrate | |
Sterile | |
Black | |
Filtration Membrane Disc |
Safety and Handling
- X300 Microsart e.motion membranes
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose