Sartorius™ Sartorius Mechatronics™ Stainless Steel Clamps


Overview
Brand: Sartorius™

Manufacturer Part Number: BBI-8854629

SS CLAMP FOR ERLEN 3000ML

UNSPSC: 41121710

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11750406

Quantity Price
1 £ 72.84 / Each
Estimated Shipment
04-04-2017
Add to basket


Description and Specification

Specification

For Use With Erlenmeyer Flasks 3000mL
Type Clamps
Material Stainless Steel
Capacity (Metric) 3000mL