Sartorius™ Sartorius Mechatronics™ Stainless Steel Clamps


Overview
Brand: Sartorius™

Manufacturer Part Number: BBI-8854564

SS CLAMP FOR FERNBACH 450ML

UNSPSC: 41121710

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11700406

Quantity Price
1 £ 38.35 / Each
Estimated Shipment
13-01-2017
Add to basket


Description and Specification

Specification

For Use With Fernbach Flasks 450mL
Type Clamps
Material Stainless Steel
Capacity (Metric) 450mL