Sartorius™ Sartorius Mechatronics™ Plastic Clamps


Overview
Brand: Sartorius™

Manufacturer Part Number: BBI-8854722

PINCE PLASTIQUE FOR ERLEN 500ML

UNSPSC: 41121710

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11700416

Quantity Price
1 £ 29.39 / Each
Estimated Shipment
06-04-2017
Add to basket


Description and Specification

Specification

For Use With Erlenmeyer Flasks 500mL
Type Clamps
Material Plastic
Capacity (Metric) 500mL