missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Hydrophobic Edged Nonsterile Cellulose Nitrate and Acetate Membrane Filters

Brand:  Sartorius™ 13507--47------N

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11700636

  • £150.00 / Pack of 100


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Hydrophobic edge membranes are used mainly for colony count and sterility testing of solutions containing substances with antibiotic characteristics. The hydrophobic edge avoids the penetration of any growth-inhibitory substance into the membrane clamp zone wherefrom it could not be rinsed out and the substance could inhibit microbial growth during incubation.

Membranes are available in three different materials:

  • Cellulose nitrate (cellulose ester), a material which assures effective retention with high flow rates and optimum colony growth
  • Cellulose acetate, a material which combines high flow rates and thermal stability with very low adsorption characteristics
  • Regenerated Cellulose, a material which combines excellent chemical and thermal stability with very low adsorption characteristics

Specifications

Specifications

0.2μm
Cellulose Acetate
White
Black
Non-sterile
47mm
For colony count; sterility testing; particle count; microscopy
Hydrophobic
Videos