Learn More
Sartorius Gridded Nonsterile Cellulose Nitrate Membrane Filters: 0.45μm Pore Size
The Nonsterile Membrane Filters are made of cellulose nitrate (cellulose mixed ester), cellulose acetate or regenerated cellulose. They can be used for colony counting, particle testing and microscopy. Choose between gridded or non-gridded, membranes, either with or without hydrophobic edge.
Brand: Sartorius 11406--47------N
Code : 53
Additional Details : Weight : 0.13000kg
Description
If agar plates or absorbent pads to be wetted with liquid culture medium are used instead of Nutrient Pad Sets, Sartorius cellulose nitrate (cellulose ester) membrane filter are recommended. Naturally, the membrane filters must be free of microbes. For this purpose, they can be boiled or autoclaved. These membranes are offered in a choice of three different colors to suit the specific test application, and provide a high-contrast background to colonies or particles.These membrane filters are in accordance with the following norms: ISO 7704, ISO 7899-2, ISO 8199, ISO 9308-1 and EN 12780. In addition to this they have been manufactured for use especially at the same time with Sartorius Nutrient Pad Sets in accordance with many international regulations and recommendations: international pharmacopoeias, DIN and ISO Standards, the American Standards for Water and Foods, mineral water regulations, brewery guidelines, such as MEBAC or EBC, and recommendations of the food industry, such as LMBG, NCA and ICUMSA. All membranes are made of cellulose nitrate (cellulose ester), a material which assures effective retention with high flow rates and optimum colony growth. The printed grid divides the filtration area into 130 squares, each measuring 3.1 x 3.1 mm makes the counting easier, especially for higher bacteria counts and for micro-colonies, but does not influence the growth.
- Outstanding recovery rates for microorganisms
- 0.45 μm are acc. to ISO 7704
- Three different colors available
- Design: 25, 47 or 50 mm in diameter, white, grey or green and gridded
- Growth Promotion Test acc. to ISO 7704: no enhancement or inhibition by the grid lines, no enhancement or inhibition due to chemical extractables
Specifications
White | |
Membrane Filter | |
Cellulose Nitrate | |
Non-sterile | |
Black | |
47 mm |
Single-use | |
Cellulose Nitrate | |
0.45 μm | |
For Colony Count, Sterility Testing, Particle Count, Microscopy | |
100 Pk. |
Safety and Handling
- nitrocellulose filters
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose