missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Extraction Thimbles

Brand:  Sartorius™ FT-1204-030100

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11771115

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

Round
Separation of particulate dust or aerosols from gas or other airstreams and emission testing at high temperature
10cm
Extraction Thimble
Glass Fiber
3cm
40
500°C
No
1.5 to 2.5mm
Videos