Learn More
Sartorius Connectors for Vivaflow™ 50
Constructed of PVC
Brand:  Sartorius VFA031
Code : NEW
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10777841
  • £38.25 / Pack of 6
In Stock
to see stock.
Add to basket
Description
Description
  • 14-558-333: T Connectors for Vivaflow™ 50 (2 pieces). For running 2 stacks of Vivaflow™ 50
  • 14-558-334: Series Interconnectors (6 pieces) for running up to six Vivaflow™ 50 units in parallel. Provides tight interconnections.
Specifications
Specifications
Series Interconnector