Learn More
Sartorius Combisart™ Accessories for Colony Counting
Accessories for Sartorius Combisart Individual and Multibranch Systems for Colony Counting
Brand: Sartorius 16841
Additional Details : Weight : 0.10000kg
Description
For use with Combisart Individual and Multibranch Systems for Colony Counting (Rep. Cat. Nos. 14-555-800 and 14-555-802).
Specifications
Base Support | |
Female Threads, TR 20 x 2 | |
Hose Nipple, DN 10 | |
ISO 8199, B.S. 304S31, AISI 304 | |
50mm Frit |
12.5cm2 | |
Stainless Steel | |
3 or 6 | |
Microbiological analysis or particle count in quality assurance |