missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Cellulose Acetate Membrane Filters

Combines high flow rates and thermal stability with very low adsorption characteristics. Sartorius Cellulose Acetate Membrane Filters are excellently suited for use in pressure filtration devices. Non-sterile filters in a variety of diameters and pore sizes are bulk-packed to save money and space.

Brand:  Sartorius 11106--30------N

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10154221

Quantity Price
1 £109.40 / Pack of 100
10 £92.29 / Pack of 100
Estimated Shipment: 24-11-2021

Add to basket


Description

Description

Combines high flow rates and thermal stability with very low adsorption characteristics. Sartorius Cellulose Acetate Membrane Filters are excellently suited for use in pressure filtration devices. Non-sterile filters in a variety of diameters and pore sizes are bulk-packed to save money and space.

  • Ideal for colony counting, particle testing, microscopy and sterility testing
  • High-quality CA membrane filters for use in pressure filtration devices
  • Available in pore sizes ranging from 0.2μm up to 5.0μm
    • 0.2μm filters are the standard for sterile filtration of aqueous solutions, such as nutrient media, buffers and sera
    • 5μm filters can be used as prefilters to retain coarse particles, while allowing microorganisms through
  • White, non-sterile filters without grids in a variety of diameters (13 to 100mm)
  • Can be sterilized by autoclaving at 121°C or 134°C; gamma irradiated to 25 kGy, dry heat or ethvlene oxide

The results of publications on adsorption are difficult to correlate, as mostly different test substances, conditions and detection methods were used, and the membranes were tested without previously being sterilized.

Specifications

Specifications

0.45 μm
Non-sterile
30 mm
Hydrophilic
For use in pressure filtration devices; Sterile filtration of solution.
Cellulose Acetate
White
120 μm
Autoclavable
100/Pk.
Videos
SDS
Documents
Certificates