missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Cellulose Acetate Membrane Filters

Combines high flow rates and thermal stability with very low adsorption characteristics. Sartorius™ Cellulose Acetate Membrane Filters are excellently suited for use in pressure filtration devices. Non-sterile filters in a variety of diameters and pore sizes are bulk-packed to save money and space.

Brand:  Sartorius™ 11107--90------G

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10528001

  • £115.00 / Pack of 25


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Ideal for colony counting, particle testing, microscopy and sterility testing
  • High-quality CA membrane filters for use in pressure filtration devices
  • Available in pore sizes ranging from 0.2μm up to 5.0μm
    • 0.2μm filters are the standard for sterile filtration of aqueous solutions, such as nutrient media, buffers and sera
    • 5μm filters can be used as prefilters to retain coarse particles, while allowing microorganisms through
  • White, non-sterile filters without grids in a variety of diameters (13 to 100mm)
  • Can be sterilized by autoclaving at 121°C or 134°C; gamma irradiated to 25 kGy, dry heat or ethvlene oxide

The results of publications on adsorption are difficult to correlate, as mostly different test substances, conditions and detection methods were used, and the membranes were tested without previously being sterilized.

Specifications

Specifications

0.2μm
90mm
No
Yes
120µm
Cellulose Acetate
White
25/pk.
For use in pressure filtration devices; Sterile filtration of solution.
Hydrophillic
Videos