Learn More
Sartorius™ Biohit PROLINE™ Plus Pipetter Starter Kits
Comfortable handle and finger support for minimized risk of strain injuries
Brand: Sartorius™ 728650
Code : 04
Additional Details : Weight : 6.12350kg
Description
Starter Kits offer an opportunity to test and get started with Proline Plus pipetters. Choose among five Starter Kits containing two or three single-channel Proline Plus pipetters. All kits include color-coded Optifit tip racks for every pipetter and a variety of useful accessories such as pipetter holders and a calibration tool, which also acts as a tube opener.
Starter Kit 1
- Two Single-Channel Pipetters (0.1-3μL, 0.5-10μL)
- Optifit tips (0.1 to 10μL), two single trays
- Two pipet holders
- Two Calibration tool/Tube openers
- Literature
- Three Single-Channel Pipetters (0.5-10μL, 10-100μL, 100-1000μL)
- Optifit tips (0.1 to 10μL, 0.5 to 200μL, 10 to 1000μL), single tray
- Three pipet holders
- Three Calibration tool/Tube openers
- Literature
- Three Single-Channel Pipetters (2-20μL, 20-200μL, 100-1000μL)
- Optifit tips (0.5 to 200μL, 10 to 1000μL), single tray
- Three pipet holders
- Three Calibration tool/Tube openers
- Literature
- Two Single-Channel Pipetters (500-5000μL, 1000-10,000μL)
- Optifit tips, bulk (100 to 5000μL, 1 to 10mL)
- Two pipet holders
- Two Calibration tool/Tube openers
- Literature
- Four Single-Channel Pipetters (0.5-10μL, 2-20μL, 20-200μL, 100-1000μL
- Optifit tips ( 0.2 to 10μL, 0.5 to 200μL, 10 to 1000μL)
- Four pipet holders
- Optifit single tray tip boxes
- Calibration tool/Tube opener
- Literature
Specifications
Finger support for minimized risk of strain injuries | |
Two single channel pipette (0.1 to 3μL, 0.5 to 10μL), optifit tip (0.1 to 10μL), two single trays, two pipette holders, two calibration tool/tube openers, literature | |
Proline™ Plus pipette starter kit 1 |