Learn More
Sartorius™ Biohit PROLINE™ Plus Pipetter Starter Kits
Comfortable handle and finger support for minimized risk of strain injuries
Brand:  Sartorius™ 728652
Code : 04
Additional Details : Weight : 0.22900kg
 View more versions of this product
Product Code. 10189859
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
Starter Kits offer an opportunity to test and get started with Proline Plus pipetters. Choose among five Starter Kits containing two or three single-channel Proline Plus pipetters. All kits include color-coded Optifit tip racks for every pipetter and a variety of useful accessories such as pipetter holders and a calibration tool, which also acts as a tube opener.
Starter Kit 1
  • Two Single-Channel Pipetters (0.1-3μL, 0.5-10μL)
  • Optifit tips (0.1 to 10μL), two single trays
  • Two pipet holders
  • Two Calibration tool/Tube openers
  • Literature
Starter Kit 2
  • Three Single-Channel Pipetters (0.5-10μL, 10-100μL, 100-1000μL)
  • Optifit tips (0.1 to 10μL, 0.5 to 200μL, 10 to 1000μL), single tray
  • Three pipet holders
  • Three Calibration tool/Tube openers
  • Literature
Starter Kit 3
  • Three Single-Channel Pipetters (2-20μL, 20-200μL, 100-1000μL)
  • Optifit tips (0.5 to 200μL, 10 to 1000μL), single tray
  • Three pipet holders
  • Three Calibration tool/Tube openers
  • Literature
Starter Kit 4
  • Two Single-Channel Pipetters (500-5000μL, 1000-10,000μL)
  • Optifit tips, bulk (100 to 5000μL, 1 to 10mL)
  • Two pipet holders
  • Two Calibration tool/Tube openers
  • Literature
Starter Kit 5
  • Four Single-Channel Pipetters (0.5-10μL, 2-20μL, 20-200μL, 100-1000μL
  • Optifit tips ( 0.2 to 10μL, 0.5 to 200μL, 10 to 1000μL)
  • Four pipet holders
  • Optifit single tray tip boxes
  • Calibration tool/Tube opener
  • Literature
Specifications
Specifications
Finger support for minimized risk of strain injuries
Three single channel pipette (2 to 20μL, 20 to 200μL, 100 to 1000μL), optifit tip (0.5 to 200μL, 10 to 1000μL), single tray, three pipette holders, three calibration
Proline™ Plus pipette starter kit 3
Videos
Documentation