Sartorius™ Biohit™ mLINE™ Pipette Multipacks
Replenish or set up a new pipetting station. Sartorius™ Biohit™ mLINE™ Pipette Multipacks conveniently combine a set of mLINE mechanical pipettes with matching tip trays and useful accessories. Available in collections of 3, 4 and 5 pipettes with matching tips
Overview
Brand: Sartorius™
Manufacturer Part Number: LH-728654
Proline Plus Starter Kit incl. 5 pipettes 0,5-10/10-100/20-200/100-1000µl
UNSPSC: 41121504
Code: NEW
Additional Details:
Additional Details: Weight: 1.20000kg
Product Code. 15154706
Quantity Price
1 £ 939.0 / 1 set
Estimated Shipment
24-02-2017
Add to basket
Description and Specification
Specification
Product Type Pipette 3+1-pack
Type Pipette 3+1-pack
Includes Pipets: .05 to 10uL, 20 to 200uL, 100 to 1000uL (one each), 8-channel 30 to 300uL, Sartorius Optifit Tip tray box: 0.2 to 10ul, 0.5 to 200ul, 5 to 350uL, 10 to 1000uL; Linear stand
For Use With (Application) Long-lasting and repetitive manual pipetting in order to prevent the risk of work related hand, arm or shoulder disorders
Includes Pipets: .05 to 10uL, 20 to 200uL, 100 to 1000uL (one each), 8-channel 30 to 300uL, Sartorius Optifit Tip tray box: 0.2 to 10ul, 0.5 to 200ul, 5 to 350uL, 10 to 1000uL; Linear stand