Learn More
Sartorius™ arium™ Comfort I Combined System (TYPE 1 - Ultrapure Water and TYPE 3 - RO Water)
Use the Sartorius™ arium™ Comfort I Combined System for producing ASTM Type 1 ultrapure water and Type 3 pure water.
Brand: Sartorius™ H2O-I-2-TOC-B
Code : NEW
Additional Details : Weight : 30.00000kg
Description
arium comfort I systems produce ASTM Type 1 ultrapure water and Type 3 pure water. The systems features the latest reverse osmosis technology, and a unique filter cartridge for producing ultrapure water of the highest quality, delivering consistently high-grade water quality at rates of up to 120l/h. The quality of this water even exceeds the ASTM Type 1 specifications. The system's integrated UV lamp (185/254 nm) prevents microbiological growth and reduces the TOC content to a minimum. The current TOC value is continuously checked by an integrated TOC monitor specially developed for this ultrapure water system and is continuously displayed.
- Reliable TOC content of <2 ppb for reproducible results
- Automatically optimised water consumption with iJust
- Easy-to-use - unique glass display with touch function
Benefits:
- Produces Type 3 and Type ASTM 1 water from tap water all in one unit; adapted to the customer's needs; the display will conveniently remain at eye level
- Optimized water consumption
- Meet customer's lab requirements and saves maintenance time
- Safe operation
- Reliable TOC reduction
- Supports reliable recording of important data
- Easy operation
- HPLC
- GC-MS, AAS, ICP-MS
- Ion chromatography
- TOC-Analysis
- Photometry
- Buffer and pH-solutions
- Feed water for ultrapure water systems, rinsing glassware and laboratory vessels, feed water for humidifiers, autoclaves and dish washer
Specifications
Type 1 to 3 | |
120 (ASTM type 1), 16 (Type 3) | |
Less than2 Product water, Less than2000 feed water | |
50.1cm | |
100/240V | |
2 to 35°C | |
Greater than99% | |
Less than1 | |
Feed water for ultrapure water system, rinsing Glassware and laboratory vessels, feed water for humidifiers, autoclaves and dish washer | |
80% | |
4 to 10 | |
Less than1/mL | |
Up to 2L/min. (ASTM type 1), Less than3L/min. (Type 3) | |
Adsorption by spherical activated Carbon, catalyst, reverse osmosis, deionization, UV radiation, optional final particle and sterile filtration | |
31kg |
18.2 Product water | |
16 | |
Less than1CFU/1000mL | |
43.5cm | |
50/60Hz | |
Greater than99% | |
47.6cm | |
5 to 45°C | |
100/240V 50/60Hz | |
2 to 30°C | |
TOC monitor | |
Wall | |
0.055 Product water, Less than1500 feed water | |
Less than4 | |
Arium™ Comfort I combined system |