Learn More
Sartorius Accessories for Biosart™ 100 Membrane Filtration
Biosart™ 100 Membrane Filtration is designed specifically for microbiological testing of pharmaceuticals, food, beverages, water and other liquids.
Brand: Sartorius 16417
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Accessories Descriptions
Adapter, ABS copolymer - 14-555-780- Biosart 100 Membrane Lifter
- For easy transfer of the membrane onto agar
- Biosart 100 Adapter
- Fits Biosart 100 Monitor onto 50 mm supports
- Biosart 100 Adapter
- 50 mm stainless-steel frit and silicone gasket
- Fits Biosart 100 Monitor onto 16840 or 16841 base
- Biosart 100 Adapter to accommodate various funnel types
- Fits Biosart 100 Monitors onto Combisart™ manifolds 16844, 16842 and 16843
- Replacement stopper for 16385
- 16385 adapts Biosart 100 Monitors on Combisart manifold
Specifications
Reusable | |
ABS Copolymer | |
For easy transfer of the membrane onto agar |
Sartorius Biosart 100 | |
Non-sterile | |
Adapter |