missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Quantitative Grade 388 Filter Papers

Brand:  Sartorius™ FT-3-101-047

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10692712

  • £8.00 / Pack of 100


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Ash-free (ash content ≤ 0.01 % according to DIN 540370)
  • Wet-strengthened
  • Color-coded box for easy selection
Specifications

Specifications

100 per pack
Disc
10 to 15μm
0.21mm
388
47mm
Black
Videos