Learn More
Sartorius Qualitative Grade 37/N Filter Papers
This paper grade is generally used for analytical and technical purposes. It has a fast flow rate due to its structure.
Brand: Sartorius FT-3-480-150
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Made of refined pulp and cotton linters with an > 95% α-cellulose content. Ash content between 0.1 - 0.15%. Wet-strengthened.Specifications
Disc | |
37/N | |
0.5 mm |
4 sec. | |
50/Pk. | |
150 mm |