Learn More
Sartorius Qualitative Grade 292a Filter Papers
These filter papers are especially suitable for qualitative analyses that require a low ash content.
Brand: Sartorius FT-4-215-150
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Pure cotton linters or cotton linters with refined pulp
- No additives, such as wet-strengthening agents
Specifications
Folded | |
97 sec. | |
100/Pk. | |
150 mm |
4 to 7 μm | |
292 a | |
0.19 mm |