Learn More
Sartorius Microsart™ Media
Enhance the safety of microbiological quality control in the pharmaceutical and biotech industries for microbial limits testing according to the USP and EP
£575.00
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15952510
|
Sartorius
14322--47----ACN |
R2A; Target microorganisms: total count, stressed organisms found in pure water systems |
£575.00
Pack of 100 |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932510
|
Sartorius
14313--47----ACN |
TSA; Target microorganisms: total count |
£575.00
Pack of 100 |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942510
|
Sartorius
14314--47----ACN |
SDA; Target microorganisms: yeasts and molds |
£575.00
Pack of 100 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Ready-to-use agar media dish (prefilled with different agar media types) with active lid
- Sterile packaged and ready to use together with Microsart filter units
- Active lid enables touch-free transfer of membrane, without using any tweezers and reduces the risk of secondary contamination
- Liftable interior lid provides easy access to select colonies after incubation for further analysis
Specifications
Growth Medium |