Sartorius™ Sartopore™ 2 MidiCaps Polypropylene Housing Polyethersulfone Membrane Filter Capsules


Overview
Brand: Sartorius™

Manufacturer Part Number: 5445307H7--OO--A

X4 SARTOPORE 2 GAMMA 0,2µ TAILLE7

UNSPSC: 41120000

Code: NEW

Additional Details:
Additional Details: Weight: 0.80000kg



Product Code. 11741585

Quantity Price
1 £ 391.2 / Pack of 4
Estimated Shipment
06-03-2017
Add to basket


Description and Specification

Specification

Filtration Area (Metric) 0.05m2
No. per Pack 4Pack
Membrane Polyethersulfone
Diameter (Metric) Outer 77mm
Pore Size 0.2μm
Autoclavable Yes
Outlet Style 0.5 in. Hose Barb
Sterile Yes
Product Type Filter Capsules
Quantity 4Pack
Housing Material Polypropylene
For Use With (Application) For therapeutics, biological fluids, injectables, media, buffers, chemicals, cleaning and sanitizing agents filtering applications
Length (Metric) 115mm
Inlet Style 0.5 in. Hose Barb
Certifications/Compliance ISO 9001, USP Class VI, cGMP Standard, HIMA and ASTM F-838-05