missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Sartopore™ 2 MidiCaps Polypropylene Housing Polyethersulfone Membrane Filter Capsules

Brand:  Sartorius™ 5445307H7

Code : NEW

Additional Details : Weight : 0.50000kg

Product Code. 15269109

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

77mm
Polypropylene
Yes
ISO 9001, USP Class VI, cGMP Standard, HIMA and ASTM F-838-05
330mm
2Pack
1.5 in. Tri-Clamp
Polyethersulfone
0.2μm
Yes
2Pack
For therapeutics, biological fluids, injectables, media, buffers, chemicals, cleaning and sanitizing agents filtering applications
0.45m2
1.5 in. Tri-Clamp
Filter Capsules
Videos