Sartorius™ Sartopore™2 Capsule 0.2μm Liquid Filters

Ensures highly economic and safe filtration performance. Sartorius™ Sartopore™2 Capsule 0.2μm Liquid Filters provide high-performance polyethersulfone (PES) liquid filtration from single-layer high flow solutions to specific Mycoplasma retentive filters. Available in a broad variety of different pore size combinations.

Overview
Brand: Sartorius™

Manufacturer Part Number: 5441307H4G-OO--B

X5 Sartopore 2 Gamma-Capsule, Poren- größe:0,45 µm/0,2 µm absolut

UNSPSC: 41000000

Code: NEW

Additional Details:
Additional Details: Weight: 0.10000kg



Product Code. 10198481

Quantity Price
1 £ 240.24 / Pack of 5
Estimated Shipment
02-03-2017
Add to basket


Description and Specification

Specification

Inlet Style 6.35mm Multiple Stepped Hose Barb
Outlet Style 6.35mm Multiple Stepped Hose Barb
Quantity 5Pack
Diameter (Metric) Outer 63mm
Material (Membrane) Polyethersulfone (PES), asymmetric
Type Gamma irradiated
Filtration Area (Metric) 0.015m2
Membrane Double layer Polyethersulfone
Pore Size 0.2μm
Autoclavable Yes
Sterile Yes
Product Type Filter Capsules
Housing Material Polypropylene
For Use With (Application) For filtration of biologicals, pharmaceuticals, cell culture media and components and buffers
Certifications/Compliance ISO 9001, USP Class VI

High-performance polyethersulfone (PES) liquid filters ensure highly economic and safe filtration performance. From single-layer high flow solutions to specific Mycoplasma retentive filters, Sartopore™2 perfectly adapts to your specific process needs. Available in a broad variety of different pore size combinations