Sartorius™ Sartobind™ Polypropylene Housing Stabilized Reinforced Cellulose Membrane S SingleSep Nano Capsules
Overview
Brand: Sartorius™
Manufacturer Part Number: 92IEXS42DN-11
SARTOBIND S SINGLESEP NANO
UNSPSC: 41120000
Code: NEW
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11790316
Quantity Price
1 £ 236.87 / Each
Estimated Shipment
01-03-2017
Add to basket
Description and Specification
Specification
Filtration Area (Metric) 36cm2
No. per Pack 1Each
Membrane Strong cation exchanger Reinforced Cellulose membrane
Pore Size 3 to 5μm
Product Type S SingleSep Nano Capsules
Quantity 1Each
Housing Material Polypropylene
For Use With (Application) For DNA, host cell proteins, endotoxins, viruses contaminant removal applications
Certifications/Compliance USP Class VI