Learn More
Sartorius™ Sartobind™ Polypropylene Housing Stabilized Reinforced Cellulose Membrane Q SingleSep Nano Capsules
Brand: Sartorius™ 92IEXQ42DN-11
Code : NEW
Additional Details : Weight : 0.01000kg
Specifications
USP Class VI | |
3 to 5 μm | |
For DNA, host cell proteins, endotoxins, viruses contaminant removal applications | |
1Each | |
Q SingleSep Nano Capsules |
Anion Exchanger Stabilized Reinforced Cellulose | |
36 cm2 | |
Polypropylene | |
1 Each |