Sartorius™ Sartobind™ Polypropylene Housing Stabilized Reinforced Cellulose Membrane Q SingleSep Mini Capsules
Overview
Brand: Sartorius™
Manufacturer Part Number: 92IEXQ42D4-OO--A
X4 SARTOBIND Q SINGLESEP MINI
UNSPSC: 41120000
Code: NEW
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 11780885
Quantity Price
1 £ 839.8 / Pack of 4
Estimated Shipment
07-04-2017
Add to basket
Description and Specification
Specification
Filtration Area (Metric) 250cm2
No. per Pack 4Pack
Membrane Anion exchanger stabilized Reinforced Cellulose membrane
Pore Size 3 to 5μm
Product Type Q SingleSep Mini Capsules
Quantity 4Pack
Housing Material Polypropylene
For Use With (Application) For DNA, host cell proteins, endotoxins, viruses contaminant removal applications
Certifications/Compliance USP Class VI