missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17258-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes the mitochondrial seryl-tRNA synthetase precursor, a member of the class II tRNA synthetase family. The mature enzyme catalyzes the ligation of Serine to tRNA(Ser) and participates in the biosynthesis of selenocysteinyl-tRNA(sec) in mitochondria. The enzyme contains an N-terminal tRNA binding domain and a core catalytic domain. It functions in a homodimeric form, which is stabilized by tRNA binding. This gene is regulated by a bidirectional promoter that also controls the expression of mitochondrial ribosomal protein S12. Both genes are within the critical interval for the autosomal domit deafness locus DFNA4 and might be linked to this disease. Multiple transcript variants encoding different isoforms have been identified for this gene.Specifications
| SARS2 | |
| Polyclonal | |
| Unconjugated | |
| SARS2 | |
| mtSerRS, SARS, SARS2, SARSM, Serine tRNA ligase, SerRS, SerRSmt, SERS, Seryl tRNA(Ser/Sec) synthetase, SYS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 292759, 54938, 71984 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9JJL8, Q9NP81 | |
| SARS2 | |
| SARS2 Fusion Protein Ag10913 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction