Learn More
Tocris Bioscience™ Sarpogrelate hydrochloride
Selective 5-HT2A antagonist
Brand:  Tocris Bioscience™ 373910
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13453524
  • £195.51 / 10mg
Estimated Shipment date
from Supplier 01-04-2020
Description
Description
Selective 5-HT2A receptor antagonist (pKi values are 8.52, 7.43 and 6.57 for 5-HT2A, 5-HT2C and 5-HT2B receptors respectively). Displays selectivity over 5-HT1, 5-HT3, 5-HT4 H1, H2, M3, α1-adrenergic, α2-adrenergic and β-adrenergic receptors. Displays cardioprotective activity in vivo.
Specifications
Specifications
Sarpogrelate hydrochloride
>99
10mg
Sarpogrelate hydrochloride
465.97