missing translation for 'onlineSavingsMsg'
Learn More

Sarcosine methyl ester, hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number p-3770911
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13515-93-0
C4H10ClNO2
139.58
MFCD00038876
HQZMRJBVCVYVQA-UHFFFAOYSA-N
sarcosine methyl ester hydrochloride, methyl 2-methylamino acetate hydrochloride, h-sar-ome.hcl, glycine, n-methyl-, methyl ester, hydrochloride, unii-ypw7yyp04f, ypw7yyp04f, sarcosine methyl ester hcl, methyl n-methylaminoacetate hydrochloride, glycine, n-methyl-, methyl ester, hydrochloride 1:1, sar-ome hcl
83544
methyl 2-(methylamino)acetate;hydrochloride
[H+].[Cl-].CNCC(=O)OC
This item is not returnable. View return policy
13515-93-0
C4H10ClNO2
139.58
MFCD00038876
HQZMRJBVCVYVQA-UHFFFAOYSA-N
sarcosine methyl ester hydrochloride, methyl 2-methylamino acetate hydrochloride, h-sar-ome.hcl, glycine, n-methyl-, methyl ester, hydrochloride, unii-ypw7yyp04f, ypw7yyp04f, sarcosine methyl ester hcl, methyl n-methylaminoacetate hydrochloride, glycine, n-methyl-, methyl ester, hydrochloride 1:1, sar-ome hcl
83544
methyl 2-(methylamino)acetate;hydrochloride
[H+].[Cl-].CNCC(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 110.0°C to 125.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 50 g
Formula Weight 139.58
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Sarcosine methyl ester, hydrochloride
compliance-icons
Product Identifier
  • Sarcosine methyl ester, hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'