missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sarcosine ethyl ester hydrochloride, 97%

Catalog Number p-3758750
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
52605-49-9
C5H12ClNO2
153.61
MFCD00012506
NIDZUMSLERGAON-UHFFFAOYSA-N
sarcosine ethyl ester hydrochloride, ethyl sarcosinate hydrochloride, ethyl 2-methylamino acetate hydrochloride, h-sar-oet.hcl, ethyl n-methylaminoacetate hydrochloride, me-gly-oet.hcl, n-methylglycine ethyl ester hydrochloride, glycine, n-methyl-, ethyl ester, hydrochloride, ethyl n-methylglycinate hydrochloride, n-methyl glycine ethyl ester hydrochloride
171173
[H+].[Cl-].CCOC(=O)CNC
This item is not returnable. View return policy
52605-49-9
C5H12ClNO2
153.61
MFCD00012506
NIDZUMSLERGAON-UHFFFAOYSA-N
sarcosine ethyl ester hydrochloride, ethyl sarcosinate hydrochloride, ethyl 2-methylamino acetate hydrochloride, h-sar-oet.hcl, ethyl n-methylaminoacetate hydrochloride, me-gly-oet.hcl, n-methylglycine ethyl ester hydrochloride, glycine, n-methyl-, ethyl ester, hydrochloride, ethyl n-methylglycinate hydrochloride, n-methyl glycine ethyl ester hydrochloride
171173
[H+].[Cl-].CCOC(=O)CNC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 123°C to 129°C
Color White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3NHCH2CO2C2H5·HCl
Quantity 10 g
IUPAC Name ethyl 2-(methylamino)acetate;hydrochloride
Formula Weight 153.61
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Sarcosine ethyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'