missing translation for 'onlineSavingsMsg'
Learn More

Saquinavir mesylate, Tocris Bioscience™

HIV protease inhibitor

Brand:  RD Systems (Bio-Techne) 4418/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13444724

  • £257.41 / 50mg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of human immunodeficiency virus (HIV) protease (Ki values are <0.1 and 0.12 nM for HIV-2 and HIV-1 protease respectively). Exhibits high antiviral activity and low cytotoxicity.
Specifications

Chemical Identifiers

149845-06-7
766.955
saquinavir mesylate, invirase, fortovase, saquinavir mesilate, saquinavir, mesylate, unii-uhb9z3841a, saquinavir monomethanesulfonate salt, saquinavir mesylate aids initiative, invirase tn, dsstox_cid_3835
CHEBI:32121
CC(C)(C)NC(=O)C1CC2CCCCC2CN1CC(C(CC3=CC=CC=C3)NC(=O)C(CC(=O)N)NC(=O)C4=NC5=CC=CC=C5C=C4)O.CS(=O)(=O)O
C39H54N6O8S
IRHXGOXEBNJUSN-YOXDLBRISA-N
60934
(2S)-N-[(2S,3R)-4-[(3S,4aS,8aS)-3-(tert-butylcarbamoyl)-3,4,4a,5,6,7,8,8a-octahydro-1H-isoquinolin-2-yl]-3-hydroxy-1-phenylbutan-2-yl]-2-(quinoline-2-carbonylamino)butanediamide;methanesulfonic acid

Specifications

Saquinavir mesylate
149845-06-7
saquinavir mesylate, invirase, fortovase, saquinavir mesilate, saquinavir, mesylate, unii-uhb9z3841a, saquinavir monomethanesulfonate salt, saquinavir mesylate aids initiative, invirase tn, dsstox_cid_3835
CC(C)(C)NC(=O)C1CC2CCCCC2CN1CC(C(CC3=CC=CC=C3)NC(=O)C(CC(=O)N)NC(=O)C4=NC5=CC=CC=C5C=C4)O.CS(=O)(=O)O
766.955
CHEBI:32121
>99%
50mg
C39H54N6O8S
IRHXGOXEBNJUSN-YOXDLBRISA-N
(2S)-N-[(2S,3R)-4-[(3S,4aS,8aS)-3-(tert-butylcarbamoyl)-3,4,4a,5,6,7,8,8a-octahydro-1H-isoquinolin-2-yl]-3-hydroxy-1-phenylbutan-2-yl]-2-(quinoline-2-carbonylamino)butanediamide;methanesulfonic acid
60934
766.95
Videos