Learn More
SAPAP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556438
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: HSKRSKSKER KPEGKPRPGM SSWWSSDDNL DSDSTYRTPS VLNRHHLGPV AHCYPDALQS PFGDLSLKTS KSNNDVKCSA CEGLALTPDA KYLKRSSWST Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.
SAPAP2 Polyclonal antibody specifically detects SAPAP2 in Human samples. It is validated for Immunohistochemistry.Specifications
SAPAP2 | |
Polyclonal | |
Unconjugated | |
DLGAP2 | |
6430596N04Rik; Dap2; DAP-2; discs large homolog associated protein 2; discs large homolog-associated protein 2; discs large-associated protein 2; discs, large (Drosophila) homolog-associated protein 2; disks large-associated protein 2; DLG associated protein 2; Dlgap2; postsynaptic density 95; PSD-95/SAP90-associated protein-2; PSD-95/SAP90-binding protein 2; SAP90/PSD-95 associated protein 2; SAP90/PSD-95-associated protein 2; SAPAP2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9228 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P1A6 | |
DLGAP2 | |
Recombinant protein corresponding to Human SAPAP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only