Learn More
Tocris Bioscience™ SANT-2
Inhibitor of Hedgehog (Hh) signaling; antagonizes smoothened activity
Brand:  Tocris Bioscience™ 361710
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13453294
  • £155.82 / 10mg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
Inhibitor of Sonic hedgehog (Shh) signaling; antagonizes smoothened receptor activity (KD = 12 nM). Displays allosteric binding characteristics similar to SANT-1 (Cat. No. 1974). Displaces smo-[3H]SAG-1.3 and -[3H]Cyclopamine binding (Ki values are 7.8 nM and 8.4 nM respectively).
Specifications
Specifications
SANT-2
>98
10mg
SANT-2
479.96