Learn More
Sanguinarine chloride, Tocris Bioscience™
Inhibitor of protein phosphatase 2C (PP2C)
Brand:  Tocris (Bio-Techne) 230210
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13430984
  • £145.53 / 10mg
Estimated Shipment date
from Supplier 23-03-2020
Description
Description
Inhibitor of protein phosphatase 2C (PP2C), selective over PP1, PP2A and PP2B in vitro. Also inhibits mitogen-activated protein kinase phosphatase-1 (MKP-1) (IC50 = 10μM). Plant alkaloid that exhibits anti-inflammatory and antioxidant activity. Blocks cell cycle in the G1 phase; inhibits cell growth in human prostate carcinoma cells.
Specifications
Chemical Identifiers
5578-73-4
367.785
sanguinarine chloride, sanguinarium chloride, viadent, sanguinarine chloride hydrate, sanguinarine hydrochloride, unii-b8z8j4400h, sanguinarine, chloride, sanguinarium chloride usan:inn, 13-methyl 1,3 benzodioxolo 5,6-c-1,3-dioxolo 4,5-i phenanthridinium chloride
C[N+]1=C2C(=C3C=CC4=C(C3=C1)OCO4)C=CC5=CC6=C(C=C52)OCO6.[Cl-]
C20H14ClNO4
GIZKAXHWLRYMLE-UHFFFAOYSA-M
68635
Specifications
10mg
5578-73-4
sanguinarine chloride, sanguinarium chloride, viadent, sanguinarine chloride hydrate, sanguinarine hydrochloride, unii-b8z8j4400h, sanguinarine, chloride, sanguinarium chloride usan:inn, 13-methyl 1,3 benzodioxolo 5,6-c-1,3-dioxolo 4,5-i phenanthridinium chloride
C[N+]1=C2C(=C3C=CC4=C(C3=C1)OCO4)C=CC5=CC6=C(C=C52)OCO6.[Cl-]
68635
>97%
Sanguinarine chloride
C20H14ClNO4
GIZKAXHWLRYMLE-UHFFFAOYSA-M
367.785
367.78