missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sand, Extra Pure, SLR, Ottawa, 20-30 Mesh, Fisher Chemical™
65.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
1 kg
3 kg
Description
Sand, Extra Pure, SLR, Ottawa, 20-30 Mesh
Specifications
Boiling Point | 2230°C |
Melting Point | 1610°C |
Color | Beige |
Physical Form | Solid |
Quantity | 1 kg |
Formula Weight | 60.08g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'