missing translation for 'onlineSavingsMsg'
Learn More

Samarium powder, -40 mesh, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 15441027
Click to view available options
Quantity:
1 g
10 g
50 g
7440-19-9
Sm
150.36
MFCD00011233 MFCD00151299
KZUNJOHGWZRPMI-UHFFFAOYSA-N
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
23951
CHEBI:33374
samarium
[Sm]
7440-19-9
Sm
150.36
MFCD00011233 MFCD00151299
KZUNJOHGWZRPMI-UHFFFAOYSA-N
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
23951
CHEBI:33374
samarium
[Sm]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Color Silver
UN Number UN3089
Quantity 10 g
Percent Purity 99.9%
Mesh Size −40 Mesh
Odor Odorless
Packaging Ampule
Sensitivity Air and moisture sensitive
Form Powder
Assay (REO)
Chemical Name or Material Samarium powder
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'